Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNNC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003279.2 | NP_003270.1 | 2, 3 | 118 | ||
XM_011529031.2 | XP_011527333.1 | 2, 3 | 128 | |||
GenBank mRNA | AK291323.1 | - | 2, 3 | 116 | ||
AK300665.1 | - | 116 | ||||
BC005323.1 | - | 2, 3 | 116 | 3934188 | ||
BU854061.1 | - | 2, 3 | 149 | |||
CR541864.1 | - | 1, 2 | 52 | |||
CR541884.1 | - | 1, 2 | 52 | |||
M22307.1 | - | 2, 3 | 99 | |||
X07898.1 | - | 2, 3 | 116 |