Search Thermo Fisher Scientific
Gene Symbol: Nek8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105804.1 | NP_001099274.1 | 3 | 466 | ||
XM_006246933.2 | XP_006246995.1 | 3 | 2382 | |||
XM_008767958.1 | XP_008766180.1 | 3 | 2382 |