Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nek8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105804.1 | NP_001099274.1 | 11 | 1593 | ||
XM_006246933.2 | XP_006246995.1 | 11 | 3509 | |||
XM_008767958.1 | XP_008766180.1 | 11 | 3509 |