Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1499 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000031.1 | NP_001000031.1 | 1 | 209 |
Gene Symbol: LOC100909578 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750853.1 | XP_003750901.1 | 1 | 209 |
Gene Symbol: Olr1496 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000716.1 | NP_001000716.1 | 1 | 209 | ||
GenBank mRNA | M64385.1 | - | 1 | 209 |