Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFAIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006291.2 | NP_006282.2 | 11 | 3009 | ||
XM_006720243.3 | XP_006720306.1 | 14 | 3193 | |||
XM_011537112.2 | XP_011535414.1 | 15 | 3325 | |||
XM_011537113.2 | XP_011535415.1 | 14 | 2725 | |||
XM_011537114.2 | XP_011535416.1 | 13 | 3651 | |||
XM_017021616.1 | XP_016877105.1 | 14 | 2720 | |||
GenBank mRNA | M92357.1 | - | 11 | 3009 |