Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008280.1 | NP_001008281.1 | 4, 5 | 639 | ||
GenBank mRNA | BC086530.1 | - | 4, 5 | 639 | 7304726 | |
D13623.1 | - | 4, 5 | 676 |