Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008281.1 | NP_001008282.1 | 7, 8 | 1229 | ||
XM_006247272.1 | XP_006247334.1 | 7, 8 | 1444 | |||
GenBank mRNA | BC085881.1 | - | 7, 8 | 1229 | 7191926 |