Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199489.4 | NP_955783.1 | 3 | 992 | ||
GenBank mRNA | AY379972.1 | - | 3 | 995 | ||
BC089762.1 | - | 979 | 7381637 |
Set Membership: |
![]() |