Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wnk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175579.3 | NP_783169.2 | 4 | 1166 | ||
XM_006247278.2 | XP_006247340.1 | 4 | 1271 | |||
XM_006247279.2 | XP_006247341.1 | 4 | 1271 | |||
XM_006247280.2 | XP_006247342.1 | 5 | 678 | |||
XM_006247281.2 | XP_006247343.1 | 3 | 430 | |||
GenBank mRNA | AY187039.1 | - | 4 | 1143 | ||
AY192567.1 | - | 4 | 1166 |