Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNT2 SIRNA Assays ›
Gene Symbol:
TNNT2
Gene Name:
troponin T2, cardiac type
Gene Aliases:
CMD1D, CMH2, CMPD2, LVNC6, RCM3, TnTC, cTnT
Chromosome Location:
Chr.1: 201359008 - 201377828 on Build GRCh38
UniGene ID:
Hs.533613
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14286
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TNNT2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000364.3
NP_000355.2
15
915
NM_001001430.2
NP_001001430.1
15
894
NM_001001431.2
NP_001001431.1
14
885
NM_001001432.2
NP_001001432.1
14
876
NM_001276345.1
NP_001263274.1
16
924
NM_001276346.1
NP_001263275.1
14
795
NM_001276347.1
NP_001263276.1
15
1027
XM_006711508.3
XP_006711571.1
15
924
XM_006711509.3
XP_006711572.1
15
1019
XM_011509938.2
XP_011508240.1
16
956
XM_011509939.1
XP_011508241.1
16
968
XM_011509940.2
XP_011508242.1
16
969
XM_011509941.2
XP_011508243.1
16
966
XM_011509942.2
XP_011508244.1
14
926
XM_011509943.2
XP_011508245.1
14
910
XM_011509944.2
XP_011508246.1
14
923
XM_011509946.1
XP_011508248.1
15
997
XM_017002216.1
XP_016857705.1
15
899
XM_017002217.1
XP_016857706.1
14
1078
GenBank mRNA
AK055533.1
-
5
1950
AK125236.1
-
14
2041
AK290621.1
-
15
1089
AY277394.1
-
12
828
BC002653.2
-
14
851
3612968
L40162.1
-
15
855
S64668.1
-
14
814
S71126.1
-
1
34
X74819.1
-
14
852
X83743.1
-
5
272
X83744.1
-
4
269
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of heart contraction
muscle filament sliding
negative regulation of ATPase activity
positive regulation of ATPase activity
regulation of muscle filament sliding speed
protein heterooligomerization
response to calcium ion
actin crosslink formation
ventricular cardiac muscle tissue morphogenesis
cardiac muscle contraction
actin binding
protein binding
tropomyosin binding
troponin C binding
protein binding, bridging
troponin I binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0