Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprc5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191591.1 | NP_001178520.1 | 3 | 1172 | ||
XM_006247678.2 | XP_006247740.1 | 3 | 2181 | |||
XM_006247679.2 | XP_006247741.1 | 3 | 2181 | |||
XM_006247681.2 | XP_006247743.1 | 3 | 1355 | |||
XM_006247682.2 | XP_006247744.1 | 3 | 1183 | |||
XM_006247683.1 | XP_006247745.1 | 3 | 1390 | |||
XM_006247684.2 | XP_006247746.1 | 3 | 1606 | |||
XM_006247685.2 | XP_006247747.1 | 3 | 1185 | |||
GenBank mRNA | BC105781.1 | - | 3 | 1109 | ||
BC168227.1 | - | 3 | 1719 | 7372263 |
Set Membership: |
![]() |