Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2v2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183052.2 | NP_898875.1 | 4 | 345 | ||
XM_006248727.2 | XP_006248789.1 | 4 | 379 | |||
GenBank mRNA | AJ515244.1 | - | 4 | 333 | ||
BC087593.1 | - | 4 | 323 | 7310168 | ||
FM082155.1 | - | 4 | 335 | |||
FQ216277.1 | - | 4 | 326 |