Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134789.1 | NP_001128261.1 | 1, 2 | 169 | ||
GenBank mRNA | CB699618.1 | - | 1, 2 | 164 | ||
CB758009.1 | - | 1, 2 | 182 | |||
CF113682.1 | - | 1, 2 | 142 | |||
FQ222889.1 | - | 1, 2 | 167 |
Gene Symbol: LOC100912534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751083.2 | XP_003751131.1 | 1, 2 | 183 |