Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrasls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105871.1 | NP_001099341.1 | 2, 3 | 300 | ||
XM_006248510.2 | XP_006248572.1 | 2, 3 | 279 | |||
GenBank mRNA | AB510983.1 | - | 1, 2 | 150 |