Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198726.2 | NP_942021.2 | 12, 13 | 1432 | ||
XM_008768713.1 | XP_008766935.1 | 13, 14 | 1632 | |||
XM_008768714.1 | XP_008766936.1 | 10, 11 | 1434 | |||
GenBank mRNA | AY351984.1 | - | 12, 13 | 1501 | ||
AY779025.1 | - | 12, 13 | 1432 |