Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025252.2 | NP_001020423.1 | 3 | 726 | ||
NM_001025253.2 | NP_001020424.1 | 3 | 444 | |||
NM_001287140.1 | NP_001274069.1 | 3 | 726 | |||
NM_001287142.1 | NP_001274071.1 | 3 | 726 | |||
NM_001287143.1 | NP_001274072.1 | 3 | 726 | |||
NM_001287144.1 | NP_001274073.1 | 3 | 444 | |||
NM_005079.3 | NP_005070.1 | 3 | 444 | |||
NR_105033.1 | 3 | 726 | ||||
NR_105034.1 | 2 | 328 | ||||
NR_105035.1 | 4 | 514 | ||||
NR_105036.1 | 3 | 389 | ||||
NR_105037.1 | 4 | 459 | ||||
GenBank mRNA | AF202897.1 | - | 3 | 670 | ||
AK057075.1 | - | 4 | 444 | |||
AK223631.1 | - | 3 | 343 | |||
AK295468.1 | - | 3 | 311 | |||
AK296615.1 | - | 3 | 448 | |||
AK308983.1 | - | 3 | 301 | |||
AK313135.1 | - | 3 | 288 | |||
BC018117.1 | - | 3 | 315 | 3916870 | ||
BI546831.1 | - | 3 | 345 | |||
BI551254.1 | - | 3 | 431 | |||
BT019444.1 | - | 3 | 228 | |||
BX640835.1 | - | 3 | 510 | |||
CR542034.1 | - | 3 | 228 | |||
CR542064.1 | - | 3 | 228 | |||
DA199944.1 | - | 4 | 326 | |||
DA409017.1 | - | 3 | 327 | |||
DA665049.1 | - | 3 | 389 | |||
DA921955.1 | - | 4 | 339 | |||
DB505575.1 | - | 2 | 226 | |||
DC331087.1 | - | 3 | 444 | |||
GQ499324.1 | - | 3 | 711 | |||
GQ499325.1 | - | 3 | 711 | |||
GQ499326.1 | - | 3 | 711 | |||
GQ499327.1 | - | 3 | 711 | |||
GQ499328.1 | - | 3 | 711 | |||
S82081.1 | - | 3 | 321 | |||
U18914.1 | - | 3 | 319 |