Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: B3galt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105887.1 | NP_001099357.1 | 3 | 736 | ||
XM_008768569.1 | XP_008766791.1 | 3 | 2791 | |||
XM_008768570.1 | XP_008766792.1 | 3 | 888 |