Gene Symbol: Olr1553 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000051.1 | NP_001000051.1 | 1 | 499 |
Gene Symbol: Olr1555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000726.1 | NP_001000726.1 | 1 | 499 |