Gene Symbol: Donson | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008287.1 | NP_001008288.1 | 8 | 1426 | ||
XM_006248047.2 | XP_006248109.1 | 8 | 1420 | |||
XM_006248048.2 | XP_006248110.1 | 8 | 1420 | |||
GenBank mRNA | BC085847.1 | - | 8 | 1426 | 7189203 |