Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006956.1 | NP_001006957.1 | 289 | |||
XM_006248824.2 | XP_006248886.1 | 575 | ||||
GenBank mRNA | BC083673.1 | - | 289 | 7135708 |
Gene Symbol: LOC100910196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248828.2 | XP_006248890.1 | 1309 | |||
XM_006248829.2 | XP_006248891.1 | 575 | ||||
XM_006248830.2 | XP_006248892.1 | 308 | ||||
XM_006248831.2 | XP_006248893.1 | 3 | 424 | |||
XM_006248832.2 | XP_006248894.1 | 243 |
Gene Symbol: LOC103690050 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751116.3 | XP_003751164.1 | 3 | 289 | ||
XM_006248816.2 | XP_006248878.1 | 3 | 575 | |||
XM_006248817.2 | XP_006248879.1 | 3 | 308 |