Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105916.1 | NP_001099386.1 | 18 | 2140 | ||
XM_006249116.2 | XP_006249178.1 | 17 | 2092 | |||
XM_006249117.1 | XP_006249179.1 | 18 | 2108 | |||
XM_006249118.1 | XP_006249180.1 | 18 | 2111 | |||
XM_008769125.1 | XP_008767347.1 | 17 | 2221 | |||
XM_008769126.1 | XP_008767348.1 | 17 | 2051 | |||
XM_008769128.1 | XP_008767350.1 | 17 | 2092 | |||
GenBank mRNA | BC166451.1 | - | 17 | 2205 | 8371649 |