Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769281.1 | XP_008767503.1 | 10 | 4436 | ||
XM_008769282.1 | XP_008767504.1 | 10 | 4436 | |||
XM_008769283.1 | XP_008767505.1 | 10 | 4435 | |||
XM_008769284.1 | XP_008767506.1 | 10 | 4435 | |||
XM_008769285.1 | XP_008767507.1 | 10 | 4435 | |||
XM_008769286.1 | XP_008767508.1 | 10 | 4435 | |||
XM_008769287.1 | XP_008767509.1 | 10 | 4436 | |||
XM_008769288.1 | XP_008767510.1 | 10 | 4435 | |||
XM_008769289.1 | XP_008767511.1 | 10 | 4436 | |||
XM_008769290.1 | XP_008767512.1 | 10 | 4435 | |||
XM_008769292.1 | XP_008767514.1 | 9 | 4226 | |||
XM_008769293.1 | XP_008767515.1 | 10 | 4435 | |||
XM_008769294.1 | XP_008767516.1 | 10 | 4436 | |||
XM_008769295.1 | XP_008767517.1 | 10 | 4436 | |||
GenBank mRNA | BC168999.1 | - | 7 | 929 |