Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249579.2 | XP_006249641.1 | 3 | 295 | ||
XM_213799.8 | XP_213799.6 | 3 | 295 |