Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr901 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000057.1 | NP_001000057.1 | 1 | 819 |
Gene Symbol: LOC103690036 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765197.1 | XP_008763419.1 | 1 | 717 |
Gene Symbol: LOC103693533 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768617.1 | XP_008766839.1 | 1 | 717 |
Gene Symbol: Olr1007 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000076.1 | NP_001000076.1 | 1 | 819 |
Gene Symbol: Olr922 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000705.1 | NP_001000705.1 | 1 | 819 |
Gene Symbol: Olr951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000073.1 | NP_001000073.1 | 1 | 819 |