Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rfx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105944.1 | NP_001099414.1 | 19 | 2838 | ||
XM_006255229.2 | XP_006255291.1 | 19 | 2990 | |||
XM_006255231.2 | XP_006255293.1 | 19 | 2990 |