Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007118.3 | NP_009049.2 | 1, 2 | 183 | ||
NR_134469.1 | 1, 2 | 183 | ||||
XM_017009801.1 | XP_016865290.1 | 1, 2 | 535 | |||
XM_017009802.1 | XP_016865291.1 | 1, 2 | 535 | |||
GenBank mRNA | AF091395.1 | - | 1, 2 | 48 | ||
BF058117.1 | - | 1, 2 | 183 | |||
U42390.1 | - | 1, 2 | 48 |