Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191670.1 | NP_001178599.1 | 9 | 2054 | ||
XM_006249937.2 | XP_006249999.1 | 10 | 2283 | |||
XM_006249938.2 | XP_006250000.1 | 10 | 2248 | |||
XM_006249939.2 | XP_006250001.1 | 10 | 2274 | |||
XM_006249940.2 | XP_006250002.1 | 10 | 2326 |