Gene Symbol: Tp53i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113775.1 | NP_001107247.1 | 5 | 394 | ||
XM_006250040.2 | XP_006250102.1 | 6 | 1177 | |||
XM_008769617.1 | XP_008767839.1 | 5 | 1262 | |||
XM_008769618.1 | XP_008767840.1 | 3 | 1105 | |||
XM_008769619.1 | XP_008767841.1 | 5 | 425 | |||
XM_008769620.1 | XP_008767842.1 | 5 | 341 | |||
XM_008769621.1 | XP_008767843.1 | 2 | 1040 | |||
GenBank mRNA | BC158771.1 | - | 6 | 441 | 7124147 |