Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tp53i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113775.1 | NP_001107247.1 | 2 | 176 | ||
XM_006250040.2 | XP_006250102.1 | 3 | 959 | |||
XM_008769617.1 | XP_008767839.1 | 2 | 1044 | |||
GenBank mRNA | BC158771.1 | - | 3 | 223 | 7124147 |