Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009631.1 | NP_001009631.1 | 231 | |||
GenBank mRNA | AF531432.1 | - | 154 | |||
BC088319.1 | - | 231 | 7325542 |