Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005534.1 | NP_001005534.1 | 3 | 129 | ||
XM_008769731.1 | XP_008767953.1 | 4 | 176 | |||
GenBank mRNA | BC082027.1 | - | 3 | 129 | 7130052 | |
FQ210665.1 | - | 3 | 131 | |||
FQ213406.1 | - | 3 | 148 | |||
FQ215668.1 | - | 4 | 183 | |||
FQ215689.1 | - | 3 | 131 | |||
FQ215712.1 | - | 3 | 131 | |||
FQ216122.1 | - | 3 | 147 | |||
FQ216612.1 | - | 3 | 148 | |||
FQ216658.1 | - | 3 | 131 | |||
FQ216892.1 | - | 3 | 147 | |||
FQ217679.1 | - | 3 | 148 | |||
FQ218940.1 | - | 3 | 148 | |||
FQ224704.1 | - | 3 | 131 | |||
FQ225195.1 | - | 3 | 142 | |||
FQ229651.1 | - | 3 | 148 | |||
FQ231805.1 | - | 3 | 148 |