Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011907.1 | NP_001011907.1 | 6 | 761 | ||
XM_003751277.2 | XP_003751325.1 | 6 | 774 | |||
GenBank mRNA | BC082067.1 | - | 6 | 761 | 7132777 | |
FQ215922.1 | - | 6 | 765 |
Gene Symbol: LOC103690046 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769779.1 | XP_008768001.1 | 3 | 392 |