Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slamf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105971.1 | NP_001099441.1 | 2 | 256 | ||
GenBank mRNA | FQ225404.1 | - | 2 | 217 | ||
FQ232537.1 | - | 2 | 217 | |||
FQ233215.1 | - | 2 | 218 | |||
FQ233573.1 | - | 2 | 217 | |||
FQ235209.1 | - | 2 | 242 |