Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TRPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012471.2 | NP_036603.1 | 9 | 3044 | ||
XM_017029774.1 | XP_016885263.1 | 10 | 3334 | |||
GenBank mRNA | AB209258.1 | - | 9 | 2458 | ||
AF054568.1 | - | 9 | 3043 | |||
BC137271.1 | - | 9 | 2884 | 9021268 | ||
BC137274.1 | - | 9 | 2884 | 9021271 |