Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105997.2 | NP_001099467.1 | 3 | 225 | ||
XM_008770009.1 | XP_008768231.1 | 4 | 592 | |||
GenBank mRNA | BC168772.1 | - | 3 | 225 | 6920150 | |
FQ217255.1 | - | 3 | 227 |
Gene Symbol: LOC100359687 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750509.3 | XP_003750557.1 | 3 | 219 |