Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ccdc158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770088.1 | XP_008768310.1 | 11 | 5957 | ||
XM_008770089.1 | XP_008768311.1 | 12 | 5225 | |||
XM_008770090.1 | XP_008768312.1 | 11 | 5945 | |||
XM_008770091.1 | XP_008768313.1 | 12 | 3138 | |||
XM_008770092.1 | XP_008768314.1 | 11 | 3068 | |||
XM_008770093.1 | XP_008768315.1 | 11 | 5957 | |||
XM_008770094.1 | XP_008768316.1 | 11 | 5957 |