Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199374.2 | NP_955406.2 | 2 | 327 | ||
XM_006250891.2 | XP_006250953.1 | 2 | 338 | |||
GenBank mRNA | AB116526.1 | - | 2 | 277 | ||
BC078996.1 | - | 2 | 327 | 7112725 |