Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013049.2 | NP_001013067.2 | 11 | 1834 | ||
XM_006251194.2 | XP_006251256.1 | 11 | 1843 | |||
XM_006251195.2 | XP_006251257.1 | 11 | 2317 | |||
XM_006251196.2 | XP_006251258.1 | 11 | 1843 | |||
XM_008770259.1 | XP_008768481.1 | 11 | 1818 |