Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003310.2 | NP_003301.1 | 7 | 900 | ||
XM_006711893.2 | XP_006711956.1 | 5 | 805 | |||
XM_011510383.2 | XP_011508685.1 | 8 | 1036 | |||
XM_011510384.2 | XP_011508686.1 | 8 | 929 | |||
XM_011510385.2 | XP_011508687.1 | 7 | 929 | |||
XM_011510387.2 | XP_011508689.1 | 6 | 691 | |||
XM_017004818.1 | XP_016860307.1 | 7 | 1001 | |||
GenBank mRNA | AF019952.1 | - | 7 | 858 | ||
AK026522.1 | - | 7 | 841 | |||
AK126288.1 | - | 10 | 1885 | |||
AK222652.1 | - | 7 | 836 | |||
BC002485.2 | - | 7 | 808 | 3349430 |