Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004210.2 | NP_001004210.1 | 2 | 265 | ||
NM_001271731.1 | NP_001258660.1 | 2 | 265 | |||
GenBank mRNA | BC079450.1 | - | 2 | 246 | 7115933 | |
BU670819.1 | - | 2 | 265 |