Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004210.2 | NP_001004210.1 | 5 | 653 | ||
NM_001271731.1 | NP_001258660.1 | 6 | 627 | |||
GenBank mRNA | BC079450.1 | - | 5 | 634 | 7115933 | |
BU670819.1 | - | 5 | 653 | |||
CB691171.1 | - | 4 | 225 |