Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106017.1 | NP_001099487.1 | 2 | 145 | ||
XM_006251216.2 | XP_006251278.1 | 3 | 245 | |||
GenBank mRNA | BC161995.1 | - | 145 | 9034230 |