Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009268.1 | NP_001009268.1 | 6 | 673 | ||
GenBank mRNA | BC088442.1 | - | 6 | 673 | 7126325 | |
FQ212547.1 | - | 6 | 672 | |||
FQ212814.1 | - | 7 | 637 | |||
FQ219071.1 | - | 6 | 622 | |||
FQ225424.1 | - | 6 | 644 | |||
FQ226083.1 | - | 6 | 619 | |||
FQ226422.1 | - | 6 | 622 | |||
FQ227142.1 | - | 6 | 622 | |||
FQ227830.1 | - | 6 | 622 | |||
FQ230292.1 | - | 2 | 165 | |||
FQ231828.1 | - | 6 | 624 | |||
FQ231874.1 | - | 6 | 657 | |||
FQ234230.1 | - | 6 | 624 | |||
FQ235071.1 | - | 6 | 644 | |||
FQ235201.1 | - | 6 | 639 |