Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009268.1 | NP_001009268.1 | 9 | 1187 | ||
GenBank mRNA | BC088442.1 | - | 9 | 1187 | 7126325 | |
FQ212547.1 | - | 9 | 1186 | |||
FQ219838.1 | - | 9 | 1153 | |||
FQ225424.1 | - | 9 | 1158 | |||
FQ226083.1 | - | 9 | 1133 | |||
FQ226422.1 | - | 9 | 1136 | |||
FQ227142.1 | - | 9 | 1136 | |||
FQ227488.1 | - | 9 | 1186 | |||
FQ227830.1 | - | 9 | 1136 | |||
FQ230292.1 | - | 5 | 679 | |||
FQ230874.1 | - | 9 | 1172 | |||
FQ231828.1 | - | 9 | 1138 | |||
FQ231874.1 | - | 9 | 1171 | |||
FQ234230.1 | - | 9 | 1138 | |||
FQ235071.1 | - | 9 | 1158 | |||
FQ235201.1 | - | 9 | 1153 |