Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024743.1 | NP_001019914.1 | 8 | 1368 | ||
XM_006251538.1 | XP_006251600.1 | 8 | 1320 | |||
GenBank mRNA | BC084711.1 | - | 3 | 475 | ||
BC097369.1 | - | 8 | 1368 | 7443949 | ||
FQ214781.1 | - | 3 | 551 |