Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC290071 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY227970.1 | - | 4 | 500 | ||
BC088274.1 | - | 4 | 522 | |||
BC088276.1 | - | 4 | 504 | 7381661 | ||
BC091200.1 | - | 4 | 556 | |||
BC091267.1 | - | 5 | 489 | 7380098 | ||
BC091273.1 | - | 4 | 666 | 7383056 | ||
BC092593.1 | - | 4 | 536 | 7379916 | ||
BC092595.1 | - | 4 | 639 | 7382388 | ||
BC092596.1 | - | 4 | 504 | 7382760 | ||
BC092597.1 | - | 4 | 493 | |||
BC093389.1 | - | 4 | 490 | 7387331 | ||
BC093392.1 | - | 4 | 541 | 7388480 | ||
DQ340291.1 | - | 4 | 488 | |||
M18853.1 | - | 4 | 469 | |||
U76836.1 | - | 4 | 407 |
Gene Symbol: LOC685078 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EX488681.1 | - | 514 |