Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogdhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106062.1 | NP_001099532.1 | 8 | 1039 | ||
XM_006252584.2 | XP_006252646.1 | 1234 |