Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HIRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003325.3 | NP_003316.3 | 18 | 2318 | ||
GenBank mRNA | AK289809.1 | - | 18 | 2098 | ||
AK299950.1 | - | 19 | 2257 | |||
AK302835.1 | - | 21 | 2544 | |||
BC039835.1 | - | 18 | 2313 | 5491361 | ||
CR456503.1 | - | 18 | 2104 | |||
X77633.1 | - | 17 | 2074 | |||
X81844.1 | - | 18 | 2159 | |||
X89887.1 | - | 18 | 2318 |