Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arglu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024972.1 | NP_001020143.1 | 4 | 921 | ||
GenBank mRNA | BC091344.1 | - | 4 | 921 | 7315932 | |
BC127459.1 | - | 4 | 878 | 7125115 |