Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coprs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169121.1 | NP_001162592.1 | 3 | 366 | ||
GenBank mRNA | CB614079.1 | - | 3 | 366 |